Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
71 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614827 MIM: 612532 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DNAJC11 - DnaJ heat shock protein family (Hsp40) member C11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018198.3 | 1509 | UTR 3 | NP_060668.2 | |||
XM_017001745.1 | 1509 | UTR 3 | XP_016857234.1 |
THAP3 - THAP domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195752.1 | 1509 | Intron | NP_001182681.1 | |||
NM_001195753.1 | 1509 | Intron | NP_001182682.1 | |||
NM_138350.3 | 1509 | UTR 3 | NP_612359.2 | |||
XM_005263532.3 | 1509 | Intron | XP_005263589.1 | |||
XM_011542400.2 | 1509 | Intron | XP_011540702.1 | |||
XM_011542401.2 | 1509 | Intron | XP_011540703.1 | |||
XM_017002761.1 | 1509 | UTR 3 | XP_016858250.1 |
Set Membership: |
HapMap JSNP Validated |