Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616879 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D22A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TBC1D22A - TBC1 domain family member 22A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284303.1 | Intron | NP_001271232.1 | ||||
NM_001284304.1 | Intron | NP_001271233.1 | ||||
NM_001284305.1 | Intron | NP_001271234.1 | ||||
NM_014346.3 | Intron | NP_055161.1 | ||||
XM_005261496.1 | Intron | XP_005261553.1 | ||||
XM_011530099.1 | Intron | XP_011528401.1 | ||||
XM_011530100.2 | Intron | XP_011528402.1 | ||||
XM_011530101.2 | Intron | XP_011528403.1 | ||||
XM_011530102.2 | Intron | XP_011528404.1 | ||||
XM_017028742.1 | Intron | XP_016884231.1 | ||||
XM_017028743.1 | Intron | XP_016884232.1 | ||||
XM_017028744.1 | Intron | XP_016884233.1 | ||||
XM_017028745.1 | Intron | XP_016884234.1 | ||||
XM_017028746.1 | Intron | XP_016884235.1 | ||||
XM_017028747.1 | Intron | XP_016884236.1 |
Set Membership: |
HapMap Validated |