Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608783 | ||||||||||||||||||||
Literature Links: |
SMYD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SMYD3 - SET and MYND domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167740.1 | 1396 | UTR 3 | NP_001161212.1 | |||
NM_022743.2 | 1396 | UTR 3 | NP_073580.1 | |||
XM_011544253.1 | 1396 | UTR 3 | XP_011542555.1 | |||
XM_011544254.2 | 1396 | UTR 3 | XP_011542556.1 | |||
XM_011544257.1 | 1396 | UTR 3 | XP_011542559.1 | |||
XM_011544258.2 | 1396 | UTR 3 | XP_011542560.1 | |||
XM_011544259.2 | 1396 | UTR 3 | XP_011542561.1 | |||
XM_011544260.1 | 1396 | UTR 3 | XP_011542562.1 | |||
XM_011544261.2 | 1396 | UTR 3 | XP_011542563.1 | |||
XM_017002094.1 | 1396 | UTR 3 | XP_016857583.1 | |||
XM_017002095.1 | 1396 | UTR 3 | XP_016857584.1 |