Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602703 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRC7 - dynein regulatory complex subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KATNB1 - katanin regulatory subunit B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005886.2 | 719 | Intron | NP_005877.2 | |||
XM_005255772.4 | 719 | Intron | XP_005255829.1 | |||
XM_006721121.3 | 719 | Intron | XP_006721184.1 | |||
XM_006721122.2 | 719 | Intron | XP_006721185.1 | |||
XM_006721123.3 | 719 | Intron | XP_006721186.1 | |||
XM_011522810.2 | 719 | UTR 5 | XP_011521112.1 | |||
XM_017022860.1 | 719 | Intron | XP_016878349.1 | |||
XM_017022861.1 | 719 | Intron | XP_016878350.1 | |||
XM_017022862.1 | 719 | UTR 5 | XP_016878351.1 | |||
XM_017022863.1 | 719 | Intron | XP_016878352.1 | |||
XM_017022864.1 | 719 | UTR 5 | XP_016878353.1 |