Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607939 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUMF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SUMF1 - sulfatase modifying factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164674.1 | 2736 | Intron | NP_001158146.1 | |||
NM_001164675.1 | 2736 | Intron | NP_001158147.1 | |||
NM_182760.3 | 2736 | Intron | NP_877437.2 | |||
XM_011533624.2 | 2736 | UTR 3 | XP_011531926.1 | |||
XM_011533625.2 | 2736 | Intron | XP_011531927.1 | |||
XM_011533626.2 | 2736 | Intron | XP_011531928.1 | |||
XM_017006252.1 | 2736 | UTR 3 | XP_016861741.1 | |||
XM_017006253.1 | 2736 | UTR 3 | XP_016861742.1 | |||
XM_017006254.1 | 2736 | Intron | XP_016861743.1 | |||
XM_017006255.1 | 2736 | Intron | XP_016861744.1 |