Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605484 | ||||||||||||||||||||
Literature Links: |
FCAMR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FCAMR - Fc fragment of IgA and IgM receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122979.2 | Intron | NP_001116451.1 | ||||
NM_001170631.1 | Intron | NP_001164102.1 | ||||
NM_032029.4 | Intron | NP_114418.2 | ||||
XM_011510051.2 | Intron | XP_011508353.1 | ||||
XM_011510052.2 | Intron | XP_011508354.1 | ||||
XM_017002442.1 | Intron | XP_016857931.1 |