Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMCN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EMCN - endomucin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159694.1 | Intron | NP_001153166.1 | ||||
NM_016242.3 | Intron | NP_057326.2 | ||||
XM_011532024.2 | Intron | XP_011530326.1 | ||||
XM_017008290.1 | Intron | XP_016863779.1 |
SNORA101A - small nucleolar RNA, H/ACA box 101A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |