Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611464 MIM: 600168 | ||||||||||||||||||||
Literature Links: |
MON1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MON1A - MON1 homolog A, secretory trafficking associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142501.1 | 2058 | Silent Mutation | CTC,CTT | L,L 482 | NP_001135973.1 | |
NM_032355.3 | 2058 | Silent Mutation | CTC,CTT | L,L 644 | NP_115731.2 | |
XM_006713345.3 | 2058 | Silent Mutation | CTC,CTT | L,L 547 | XP_006713408.1 | |
XM_011534160.1 | 2058 | Silent Mutation | CTC,CTT | L,L 547 | XP_011532462.1 |
MST1R - macrophage stimulating 1 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |