Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151523 MIM: 607972 MIM: 601729 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD37 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD37 - CD37 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040031.1 | Intron | NP_001035120.1 | ||||
NM_001774.2 | Intron | NP_001765.1 | ||||
XM_005259435.3 | Intron | XP_005259492.1 | ||||
XM_005259436.3 | Intron | XP_005259493.1 | ||||
XM_011527542.2 | Intron | XP_011525844.1 | ||||
XM_011527543.2 | Intron | XP_011525845.1 | ||||
XM_011527544.2 | Intron | XP_011525846.1 | ||||
XM_017027513.1 | Intron | XP_016883002.1 |
SLC6A16 - solute carrier family 6 member 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEAD2 - TEA domain transcription factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |