Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614130 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADAD1 - adenosine deaminase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159285.1 | Intron | NP_001152757.1 | ||||
NM_001159295.1 | Intron | NP_001152767.1 | ||||
NM_139243.3 | Intron | NP_640336.1 | ||||
XM_005262741.2 | Intron | XP_005262798.1 | ||||
XM_005262743.2 | Intron | XP_005262800.1 | ||||
XM_005262744.2 | Intron | XP_005262801.1 | ||||
XM_005262745.2 | Intron | XP_005262802.1 | ||||
XM_011531603.1 | Intron | XP_011529905.1 |
Set Membership: |
HapMap Validated |