Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNIP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNIP3 - TNFAIP3 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128843.2 | Intron | NP_001122315.2 | ||||
NM_001244764.1 | Intron | NP_001231693.1 | ||||
NM_024873.5 | Intron | NP_079149.3 | ||||
XM_011532256.2 | Intron | XP_011530558.1 | ||||
XM_011532257.2 | Intron | XP_011530559.1 | ||||
XM_017008619.1 | Intron | XP_016864108.1 | ||||
XM_017008620.1 | Intron | XP_016864109.1 | ||||
XM_017008621.1 | Intron | XP_016864110.1 | ||||
XM_017008622.1 | Intron | XP_016864111.1 | ||||
XM_017008623.1 | Intron | XP_016864112.1 | ||||
XM_017008624.1 | Intron | XP_016864113.1 | ||||
XM_017008625.1 | Intron | XP_016864114.1 |