Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606767 MIM: 609050 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNG3 - potassium voltage-gated channel modifier subfamily G member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTA3 - metastasis associated 1 family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282755.1 | Intron | NP_001269684.1 | ||||
NM_001282756.1 | Intron | NP_001269685.1 | ||||
NM_020744.3 | Intron | NP_065795.1 | ||||
XM_005264456.2 | Intron | XP_005264513.1 | ||||
XM_005264458.2 | Intron | XP_005264515.1 | ||||
XM_005264459.2 | Intron | XP_005264516.1 | ||||
XM_017004561.1 | Intron | XP_016860050.1 | ||||
XM_017004562.1 | Intron | XP_016860051.1 |
Set Membership: |
HapMap |