Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606133 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42EP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDC42EP3 - CDC42 effector protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270436.1 | 2601 | UTR 3 | NP_001257365.1 | |||
NM_001270437.1 | 2601 | UTR 3 | NP_001257366.1 | |||
NM_001270438.1 | 2601 | UTR 3 | NP_001257367.1 | |||
NM_006449.4 | 2601 | UTR 3 | NP_006440.2 | |||
XM_011532482.2 | 2601 | UTR 3 | XP_011530784.1 |