Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601618 MIM: 604784 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOX18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SOX18 - SRY-box 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCEA2 - transcription elongation factor A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003195.4 | Intron | NP_003186.1 | ||||
NM_198723.1 | Intron | NP_942016.1 | ||||
XM_005260229.3 | Intron | XP_005260286.1 | ||||
XM_011529022.1 | Intron | XP_011527324.1 | ||||
XM_011529023.1 | Intron | XP_011527325.1 | ||||
XM_011529027.2 | Intron | XP_011527329.1 | ||||
XM_017028036.1 | Intron | XP_016883525.1 | ||||
XM_017028037.1 | Intron | XP_016883526.1 |