Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610866 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UCKL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UCKL1-AS1 - UCKL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF512B - zinc finger protein 512B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020713.2 | 455 | UTR 3 | NP_065764.1 | |||
XM_005260226.3 | 455 | UTR 3 | XP_005260283.1 | |||
XM_011528929.2 | 455 | UTR 3 | XP_011527231.1 | |||
XM_011528930.2 | 455 | UTR 3 | XP_011527232.1 | |||
XM_011528931.2 | 455 | UTR 3 | XP_011527233.1 | |||
XM_011528932.2 | 455 | UTR 3 | XP_011527234.1 | |||
XM_011528933.2 | 455 | Intron | XP_011527235.1 |
Set Membership: |
HapMap |