Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118504 | ||||||||||||||||||||
Literature Links: |
CHRNA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNA4 - cholinergic receptor nicotinic alpha 4 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000744.6 | 3438 | UTR 3 | NP_000735.1 | |||
NM_001256573.1 | 3438 | UTR 3 | NP_001243502.1 | |||
XM_011528524.1 | 3438 | UTR 3 | XP_011526826.1 | |||
XM_017027624.1 | 3438 | UTR 3 | XP_016883113.1 | |||
XM_017027625.1 | 3438 | UTR 3 | XP_016883114.1 |