Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614047 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTHFD2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTHFD2L - methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144978.1 | Intron | NP_001138450.1 | ||||
XM_005265690.2 | Intron | XP_005265747.1 | ||||
XM_017008218.1 | Intron | XP_016863707.1 | ||||
XM_017008219.1 | Intron | XP_016863708.1 | ||||
XM_017008220.1 | Intron | XP_016863709.1 | ||||
XM_017008221.1 | Intron | XP_016863710.1 | ||||
XM_017008222.1 | Intron | XP_016863711.1 | ||||
XM_017008223.1 | Intron | XP_016863712.1 | ||||
XM_017008224.1 | Intron | XP_016863713.1 | ||||
XM_017008225.1 | Intron | XP_016863714.1 | ||||
XM_017008226.1 | Intron | XP_016863715.1 | ||||
XM_017008227.1 | Intron | XP_016863716.1 | ||||
XM_017008228.1 | Intron | XP_016863717.1 | ||||
XM_017008229.1 | Intron | XP_016863718.1 | ||||
XM_017008230.1 | Intron | XP_016863719.1 | ||||
XM_017008231.1 | Intron | XP_016863720.1 | ||||
XM_017008232.1 | Intron | XP_016863721.1 | ||||
XM_017008233.1 | Intron | XP_016863722.1 | ||||
XM_017008234.1 | Intron | XP_016863723.1 | ||||
XM_017008235.1 | Intron | XP_016863724.1 |