Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606715 MIM: 610405 | ||||||||||||||||||||
Literature Links: |
ASIC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASIC4 - acid sensing ion channel subunit family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182847.2 | 2460 | Intron | NP_878267.2 | |||
XM_005246671.2 | 2460 | Intron | XP_005246728.2 | |||
XM_017004439.1 | 2460 | Intron | XP_016859928.1 |
CHPF - chondroitin polymerizing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195731.1 | 2460 | UTR 3 | NP_001182660.1 | |||
NM_024536.5 | 2460 | UTR 3 | NP_078812.2 | |||
XM_011511838.2 | 2460 | UTR 3 | XP_011510140.1 |
TMEM198 - transmembrane protein 198 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |