Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615003 MIM: 610626 MIM: 607083 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDHD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDHD2 - DDHD domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLPP5 - phospholipid phosphatase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WHSC1L1 - Wolf-Hirschhorn syndrome candidate 1-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017778.2 | Intron | NP_060248.2 | ||||
NM_023034.1 | Intron | NP_075447.1 |