Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612758 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TAPT1 - transmembrane anterior posterior transformation 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153365.2 | 4550 | UTR 3 | NP_699196.2 | |||
XM_005248139.2 | 4550 | UTR 3 | XP_005248196.1 | |||
XM_005248140.3 | 4550 | UTR 3 | XP_005248197.1 | |||
XM_011513812.2 | 4550 | UTR 3 | XP_011512114.1 | |||
XM_011513815.2 | 4550 | UTR 3 | XP_011512117.1 | |||
XM_011513816.2 | 4550 | UTR 3 | XP_011512118.1 | |||
XM_011513817.2 | 4550 | UTR 3 | XP_011512119.1 | |||
XM_017007876.1 | 4550 | UTR 3 | XP_016863365.1 |