Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
51 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611889 MIM: 603623 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
S100PBP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
S100PBP - S100P binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256121.1 | Intron | NP_001243050.1 | ||||
NM_022753.3 | Intron | NP_073590.2 | ||||
XM_011541961.2 | Intron | XP_011540263.1 | ||||
XM_011541962.2 | Intron | XP_011540264.1 | ||||
XM_011541963.1 | Intron | XP_011540265.1 | ||||
XM_017002098.1 | Intron | XP_016857587.1 |
YARS - tyrosyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |