Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608304 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAGE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAGE1 - cancer antigen 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIOK1 - RIO kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031480.2 | Intron | NP_113668.2 | ||||
NM_153005.1 | Intron | NP_694550.1 | ||||
XM_005249446.3 | Intron | XP_005249503.1 | ||||
XM_011514933.2 | Intron | XP_011513235.2 |
Set Membership: |
HapMap |