Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609245 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLCC1 - chloride channel CLIC like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048210.2 | 2802 | UTR 3 | NP_001041675.1 | |||
NM_001278202.1 | 2802 | UTR 3 | NP_001265131.1 | |||
NM_001278203.1 | 2802 | UTR 3 | NP_001265132.1 | |||
NM_015127.4 | 2802 | UTR 3 | NP_055942.1 |
GPSM2 - G-protein signaling modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321038.1 | 2802 | UTR 3 | NP_001307967.1 | |||
NM_001321039.1 | 2802 | Intron | NP_001307968.1 | |||
NM_013296.4 | 2802 | UTR 3 | NP_037428.3 | |||
XM_006710589.2 | 2802 | UTR 3 | XP_006710652.1 | |||
XM_011541301.2 | 2802 | UTR 3 | XP_011539603.1 | |||
XM_011541302.2 | 2802 | UTR 3 | XP_011539604.1 | |||
XM_011541303.2 | 2802 | Intron | XP_011539605.1 | |||
XM_017001097.1 | 2802 | UTR 3 | XP_016856586.1 | |||
XM_017001098.1 | 2802 | UTR 3 | XP_016856587.1 |
Set Membership: |
HapMap |