Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600629 MIM: 170261 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-DOB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HLA-DOB - major histocompatibility complex, class II, DO beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAP2 - transporter 2, ATP binding cassette subfamily B member | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000544.3 | 4938 | UTR 3 | NP_000535.3 | |||
NM_001290043.1 | 4938 | UTR 3 | NP_001276972.1 | |||
NM_018833.2 | 4938 | Intron | NP_061313.2 |
Set Membership: |
HapMap Validated |