Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 120290 MIM: 601417 MIM: 611500 MIM: 602045 MIM: 180246 MIM: 601416 | |||||||||||||||||||||||
Literature Links: |
COL11A2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
COL11A2 - collagen type XI alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSD17B8 - hydroxysteroid 17-beta dehydrogenase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR219A1 - microRNA 219a-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RING1 - ring finger protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RXRB - retinoid X receptor beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A7 - solute carrier family 39 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077516.1 | 1259 | Missense Mutation | GAA,GGA | E,G 280 | NP_001070984.1 | |
NM_001288777.1 | 1259 | Missense Mutation | GAA,GGA | E,G 155 | NP_001275706.1 | |
NM_006979.2 | 1259 | Missense Mutation | GAA,GGA | E,G 280 | NP_008910.2 |
Set Membership: |
HapMap |