Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
61 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606168 MIM: 605411 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDX20 - DEAD-box helicase 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCND3 - potassium voltage-gated channel subfamily D member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004980.4 | 4002 | Intron | NP_004971.2 | |||
NM_172198.2 | 4002 | Intron | NP_751948.1 | |||
XM_005270851.4 | 4002 | UTR 3 | XP_005270908.1 | |||
XM_006710629.3 | 4002 | UTR 3 | XP_006710692.1 | |||
XM_006710630.3 | 4002 | UTR 3 | XP_006710693.1 | |||
XM_006710631.3 | 4002 | Intron | XP_006710694.1 | |||
XM_006710632.3 | 4002 | Intron | XP_006710695.1 | |||
XM_011541425.2 | 4002 | Intron | XP_011539727.1 | |||
XM_011541426.2 | 4002 | Intron | XP_011539728.1 | |||
XM_011541427.2 | 4002 | Intron | XP_011539729.1 | |||
XM_011541428.2 | 4002 | Intron | XP_011539730.1 | |||
XM_017001244.1 | 4002 | UTR 3 | XP_016856733.1 | |||
XM_017001245.1 | 4002 | Intron | XP_016856734.1 |
Set Membership: |
HapMap |