Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 103020 | ||||||||||||||||||||
Literature Links: |
AK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AK2 - adenylate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199199.1 | 662 | Missense Mutation | GCA,GTA | A,V 217 | NP_001186128.1 | |
NM_001319139.1 | 662 | Missense Mutation | GCA,GTA | A,V 177 | NP_001306068.1 | |
NM_001319140.1 | 662 | Missense Mutation | GCA,GTA | A,V 177 | NP_001306069.1 | |
NM_001319141.1 | 662 | Missense Mutation | GCA,GTA | A,V 225 | NP_001306070.1 | |
NM_001319142.1 | 662 | Missense Mutation | GCA,GTA | A,V 183 | NP_001306071.1 | |
NM_001319143.1 | 662 | UTR 3 | NP_001306072.1 | |||
NM_001625.3 | 662 | Missense Mutation | GCA,GTA | A,V 225 | NP_001616.1 | |
NM_013411.4 | 662 | Missense Mutation | GCA,GTA | A,V 225 | NP_037543.1 |