Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 156570 | ||||||||||||||||||||
Literature Links: |
MTR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTR - 5-methyltetrahydrofolate-homocysteine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000254.2 | Intron | NP_000245.2 | ||||
NM_001291939.1 | Intron | NP_001278868.1 | ||||
NM_001291940.1 | Intron | NP_001278869.1 | ||||
XM_005273141.4 | Intron | XP_005273198.1 | ||||
XM_006711770.2 | Intron | XP_006711833.1 | ||||
XM_011544194.2 | Intron | XP_011542496.1 | ||||
XM_017001329.1 | Intron | XP_016856818.1 | ||||
XM_017001330.1 | Intron | XP_016856819.1 |