Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611327 MIM: 603444 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB4 - DnaJ heat shock protein family (Hsp40) member B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317099.1 | 321 | Intron | NP_001304028.1 | |||
NM_001317100.1 | 321 | UTR 5 | NP_001304029.1 | |||
NM_001317101.1 | 321 | UTR 5 | NP_001304030.1 | |||
NM_001317102.1 | 321 | Intron | NP_001304031.1 | |||
NM_001317103.1 | 321 | Intron | NP_001304032.1 | |||
NM_007034.4 | 321 | Intron | NP_008965.2 |
FUBP1 - far upstream element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303433.1 | 321 | Intron | NP_001290362.1 | |||
NM_003902.4 | 321 | Intron | NP_003893.2 | |||
XM_011542391.2 | 321 | Intron | XP_011540693.1 | |||
XM_011542392.2 | 321 | Intron | XP_011540694.1 | |||
XM_011542393.2 | 321 | Intron | XP_011540695.1 | |||
XM_017002739.1 | 321 | Intron | XP_016858228.1 | |||
XM_017002740.1 | 321 | Intron | XP_016858229.1 | |||
XM_017002741.1 | 321 | UTR 5 | XP_016858230.1 | |||
XM_017002742.1 | 321 | Intron | XP_016858231.1 | |||
XM_017002743.1 | 321 | Intron | XP_016858232.1 | |||
XM_017002744.1 | 321 | Intron | XP_016858233.1 |