Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611012 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGSL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RGSL1 - regulator of G-protein signaling like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137669.1 | 99 | Intron | NP_001131141.1 | |||
XM_005245137.1 | 99 | Intron | XP_005245194.1 | |||
XM_011509494.2 | 99 | Intron | XP_011507796.1 | |||
XM_011509496.1 | 99 | Intron | XP_011507798.1 | |||
XM_011509500.1 | 99 | Intron | XP_011507802.1 | |||
XM_011509503.1 | 99 | Intron | XP_011507805.1 | |||
XM_011509504.2 | 99 | Intron | XP_011507806.1 | |||
XM_017001188.1 | 99 | Intron | XP_016856677.1 | |||
XM_017001189.1 | 99 | Intron | XP_016856678.1 | |||
XM_017001190.1 | 99 | Intron | XP_016856679.1 | |||
XM_017001191.1 | 99 | Intron | XP_016856680.1 | |||
XM_017001192.1 | 99 | UTR 5 | XP_016856681.1 | |||
XM_017001193.1 | 99 | Intron | XP_016856682.1 | |||
XM_017001194.1 | 99 | Intron | XP_016856683.1 | |||
XM_017001195.1 | 99 | Intron | XP_016856684.1 | |||
XM_017001196.1 | 99 | Intron | XP_016856685.1 | |||
XM_017001197.1 | 99 | Intron | XP_016856686.1 |