Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608803 MIM: 139270 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJC2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GJC2 - gap junction protein gamma 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GUK1 - guanylate kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000858.5 | Intron | NP_000849.1 | ||||
NM_001159390.1 | Intron | NP_001152862.1 | ||||
NM_001159391.1 | Intron | NP_001152863.1 | ||||
NM_001242839.1 | Intron | NP_001229768.1 | ||||
NM_001242840.1 | Intron | NP_001229769.1 | ||||
XM_005273103.3 | Intron | XP_005273160.1 | ||||
XM_005273104.4 | Intron | XP_005273161.1 | ||||
XM_005273105.3 | Intron | XP_005273162.1 | ||||
XM_005273106.2 | Intron | XP_005273163.1 | ||||
XM_011544174.2 | Intron | XP_011542476.1 | ||||
XM_017001091.1 | Intron | XP_016856580.1 | ||||
XM_017001092.1 | Intron | XP_016856581.1 | ||||
XM_017001093.1 | Intron | XP_016856582.1 | ||||
XM_017001094.1 | Intron | XP_016856583.1 | ||||
XM_017001095.1 | Intron | XP_016856584.1 | ||||
XM_017001096.1 | Intron | XP_016856585.1 |
Set Membership: |
HapMap |