Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf35 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C10orf35 - chromosome 10 open reading frame 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145306.2 | 819 | Silent Mutation | AAC,AAT | N,N 36 | NP_660349.1 | |
XM_005269606.1 | 819 | Silent Mutation | AAC,AAT | N,N 78 | XP_005269663.1 | |
XM_005269608.3 | 819 | Silent Mutation | AAC,AAT | N,N 36 | XP_005269665.1 | |
XM_011539455.2 | 819 | Silent Mutation | AAC,AAT | N,N 36 | XP_011537757.1 | |
XM_017015836.1 | 819 | Silent Mutation | AAC,AAT | N,N 36 | XP_016871325.1 |