Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 111300 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GYPA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GYPA - glycophorin A (MNS blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308187.1 | 2495 | UTR 3 | NP_001295116.1 | |||
NM_001308190.1 | 2495 | UTR 3 | NP_001295119.1 | |||
NM_002099.7 | 2495 | UTR 3 | NP_002090.4 | |||
XM_017008134.1 | 2495 | UTR 3 | XP_016863623.1 | |||
XM_017008135.1 | 2495 | UTR 3 | XP_016863624.1 | |||
XM_017008136.1 | 2495 | UTR 3 | XP_016863625.1 |