Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616618 MIM: 608221 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACBD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MASTL - microtubule associated serine/threonine kinase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172303.2 | 2886 | Intron | NP_001165774.1 | |||
NM_001172304.2 | 2886 | Intron | NP_001165775.1 | |||
NM_001320756.1 | 2886 | Intron | NP_001307685.1 | |||
NM_001320757.1 | 2886 | Intron | NP_001307686.1 | |||
NM_032844.4 | 2886 | Intron | NP_116233.2 | |||
XM_005252631.3 | 2886 | Intron | XP_005252688.1 | |||
XM_005252632.3 | 2886 | Intron | XP_005252689.1 | |||
XM_006717519.3 | 2886 | Intron | XP_006717582.1 | |||
XM_006717520.3 | 2886 | Intron | XP_006717583.1 | |||
XM_011519751.2 | 2886 | Intron | XP_011518053.1 | |||
XM_017016852.1 | 2886 | Intron | XP_016872341.1 | |||
XM_017016853.1 | 2886 | Intron | XP_016872342.1 | |||
XM_017016854.1 | 2886 | Intron | XP_016872343.1 | |||
XM_017016855.1 | 2886 | Intron | XP_016872344.1 | |||
XM_017016856.1 | 2886 | Intron | XP_016872345.1 |
Set Membership: |
HapMap Validated |