Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608221 MIM: 607472 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MASTL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MASTL - microtubule associated serine/threonine kinase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172303.2 | Intron | NP_001165774.1 | ||||
NM_001172304.2 | Intron | NP_001165775.1 | ||||
NM_001320756.1 | Intron | NP_001307685.1 | ||||
NM_001320757.1 | Intron | NP_001307686.1 | ||||
NM_032844.4 | Intron | NP_116233.2 | ||||
XM_005252631.3 | Intron | XP_005252688.1 | ||||
XM_005252632.3 | Intron | XP_005252689.1 | ||||
XM_006717519.3 | Intron | XP_006717582.1 | ||||
XM_006717520.3 | Intron | XP_006717583.1 | ||||
XM_011519751.2 | Intron | XP_011518053.1 | ||||
XM_017016852.1 | Intron | XP_016872341.1 | ||||
XM_017016853.1 | Intron | XP_016872342.1 | ||||
XM_017016854.1 | Intron | XP_016872343.1 | ||||
XM_017016855.1 | Intron | XP_016872344.1 | ||||
XM_017016856.1 | Intron | XP_016872345.1 |
YME1L1 - YME1 like 1 ATPase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |