Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611963 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC344967 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC344967 - acyl-CoA thioesterase 7 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
N4BP2 - NEDD4 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318359.1 | Intron | NP_001305288.1 | ||||
NM_018177.5 | Intron | NP_060647.2 | ||||
XM_006714023.3 | Intron | XP_006714086.1 | ||||
XM_011513716.2 | Intron | XP_011512018.1 | ||||
XM_011513717.2 | Intron | XP_011512019.1 | ||||
XM_011513718.2 | Intron | XP_011512020.1 | ||||
XM_011513719.2 | Intron | XP_011512021.1 | ||||
XM_011513720.1 | Intron | XP_011512022.1 | ||||
XM_011513721.1 | Intron | XP_011512023.1 | ||||
XM_017008397.1 | Intron | XP_016863886.1 | ||||
XM_017008398.1 | Intron | XP_016863887.1 | ||||
XM_017008399.1 | Intron | XP_016863888.1 |