Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602846 | ||||||||||||||||||||
Literature Links: |
UBE2K PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2K - ubiquitin conjugating enzyme E2 K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111112.1 | 159 | UTR 5 | NP_001104582.1 | |||
NM_001111113.1 | 159 | UTR 5 | NP_001104583.1 | |||
NM_001312646.1 | 159 | UTR 5 | NP_001299575.1 | |||
NM_001312647.1 | 159 | UTR 5 | NP_001299576.1 | |||
NM_001312648.1 | 159 | UTR 5 | NP_001299577.1 | |||
NM_005339.4 | 159 | UTR 5 | NP_005330.1 |