Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611135 MIM: 607031 MIM: 603686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
EAS - Not Available | African American
|
YRI (Yoruba)
|
||||||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
||||||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR - Not Available | ||||||||
AMR - Not Available |
KLB - klotho beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIAS - lipoic acid synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL9 - ribosomal protein L9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000661.4 | Intron | NP_000652.2 | ||||
NM_001024921.3 | Intron | NP_001020092.1 |
Set Membership: |
HapMap Validated |