Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606347 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSTPIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PSTPIP1 - proline-serine-threonine phosphatase interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321135.1 | Intron | NP_001308064.1 | ||||
NM_001321136.1 | Intron | NP_001308065.1 | ||||
NM_001321137.1 | Intron | NP_001308066.1 | ||||
NM_003978.4 | Intron | NP_003969.2 | ||||
XM_006720737.2 | Intron | XP_006720800.1 | ||||
XM_011522163.1 | Intron | XP_011520465.1 | ||||
XM_011522164.1 | Intron | XP_011520466.1 | ||||
XM_011522165.2 | Intron | XP_011520467.1 | ||||
XM_011522166.1 | Intron | XP_011520468.1 | ||||
XM_011522167.1 | Intron | XP_011520469.1 | ||||
XM_011522168.2 | Intron | XP_011520470.1 | ||||
XM_011522169.1 | Intron | XP_011520471.1 |
Set Membership: |
HapMap |