Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605564 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CIB2 - calcium and integrin binding family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271888.1 | 622 | Silent Mutation | GAC,GAT | D,D 116 | NP_001258817.1 | |
NM_001271889.1 | 622 | Silent Mutation | GAC,GAT | D,D 110 | NP_001258818.1 | |
NM_001301224.1 | 622 | Silent Mutation | GAC,GAT | D,D 164 | NP_001288153.1 | |
NM_006383.3 | 622 | Silent Mutation | GAC,GAT | D,D 159 | NP_006374.1 | |
XM_005254126.3 | 622 | Silent Mutation | GAC,GAT | D,D 159 | XP_005254183.1 | |
XM_006720374.2 | 622 | Silent Mutation | GAC,GAT | D,D 116 | XP_006720437.1 | |
XM_011521161.2 | 622 | Silent Mutation | GAC,GAT | D,D 116 | XP_011519463.1 |
SH2D7 - SH2 domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |