Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614362 MIM: 601149 | ||||||||||||||||||||
Literature Links: |
ACSBG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSBG1 - acyl-CoA synthetase bubblegum family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199377.1 | 2786 | UTR 3 | NP_001186306.1 | |||
NM_015162.4 | 2786 | UTR 3 | NP_055977.3 | |||
XM_011521390.2 | 2786 | UTR 3 | XP_011519692.1 | |||
XM_011521391.2 | 2786 | Intron | XP_011519693.2 | |||
XM_017022025.1 | 2786 | Intron | XP_016877514.1 | |||
XM_017022026.1 | 2786 | UTR 3 | XP_016877515.1 |
IDH3A - isocitrate dehydrogenase 3 (NAD(+)) alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005530.2 | 2786 | Intron | NP_005521.1 | |||
XM_005254336.2 | 2786 | Intron | XP_005254393.1 | |||
XM_017022118.1 | 2786 | Intron | XP_016877607.1 | |||
XM_017022119.1 | 2786 | Intron | XP_016877608.1 | |||
XM_017022120.1 | 2786 | Intron | XP_016877609.1 |