Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612501 | ||||||||||||||||||||
Literature Links: |
UBE2Q2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2Q2 - ubiquitin conjugating enzyme E2 Q2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145335.1 | Intron | NP_001138807.1 | ||||
NM_001284382.1 | Intron | NP_001271311.1 | ||||
NM_173469.3 | Intron | NP_775740.1 | ||||
XM_005254787.1 | Intron | XP_005254844.1 | ||||
XM_005254788.1 | Intron | XP_005254845.1 | ||||
XM_006720770.1 | Intron | XP_006720833.1 | ||||
XM_011522228.1 | Intron | XP_011520530.1 | ||||
XM_017022726.1 | Intron | XP_016878215.1 | ||||
XM_017022727.1 | Intron | XP_016878216.1 | ||||
XM_017022728.1 | Intron | XP_016878217.1 |