Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194539 | ||||||||||||||||||||
Literature Links: |
ZNF32 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF32 - zinc finger protein 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005368.2 | 933 | Missense Mutation | AAC,CAC | N,H 263 | NP_001005368.1 | |
NM_001324164.1 | 933 | Missense Mutation | AAC,CAC | N,H 267 | NP_001311093.1 | |
NM_001324165.1 | 933 | Missense Mutation | AAC,CAC | N,H 267 | NP_001311094.1 | |
NM_001324166.1 | 933 | Missense Mutation | AAC,CAC | N,H 238 | NP_001311095.1 | |
NM_001324167.1 | 933 | Missense Mutation | AAC,CAC | N,H 238 | NP_001311096.1 | |
NM_001324168.1 | 933 | Missense Mutation | AAC,CAC | N,H 238 | NP_001311097.1 | |
NM_006973.2 | 933 | Missense Mutation | AAC,CAC | N,H 263 | NP_008904.1 | |
XM_017016616.1 | 933 | Missense Mutation | AAC,CAC | N,H 263 | XP_016872105.1 |
ZNF32-AS1 - ZNF32 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF32-AS2 - ZNF32 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF32-AS3 - ZNF32 antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |