Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300375 MIM: 300368 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CHST7 - carbohydrate sulfotransferase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9A7 - solute carrier family 9 member A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257291.1 | 4936 | UTR 3 | NP_001244220.1 | |||
NM_032591.2 | 4936 | UTR 3 | NP_115980.1 | |||
XM_005272681.4 | 4936 | Intron | XP_005272738.1 | |||
XM_005272682.4 | 4936 | Intron | XP_005272739.1 | |||
XM_006724564.3 | 4936 | Intron | XP_006724627.1 | |||
XM_011543990.2 | 4936 | UTR 3 | XP_011542292.1 | |||
XM_017029905.1 | 4936 | UTR 3 | XP_016885394.1 | |||
XM_017029906.1 | 4936 | Intron | XP_016885395.1 |
Set Membership: |
HapMap Validated |