Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609230 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUDT5 - nudix hydrolase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321647.1 | 1025 | Silent Mutation | GCA,GCG | A,A 200 | NP_001308576.1 | |
NM_001321648.1 | 1025 | Silent Mutation | GCA,GCG | A,A 117 | NP_001308577.1 | |
NM_014142.3 | 1025 | Silent Mutation | GCA,GCG | A,A 203 | NP_054861.2 |
SEC61A2 - Sec61 translocon alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142627.2 | 1025 | Intron | NP_001136099.1 | |||
NM_001142628.1 | 1025 | Intron | NP_001136100.1 | |||
NM_018144.3 | 1025 | Intron | NP_060614.2 |
Set Membership: |
HapMap |