Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608326 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STOML1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
STOML1 - stomatin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256672.1 | 2826 | UTR 3 | NP_001243601.1 | |||
NM_001256673.1 | 2826 | UTR 3 | NP_001243602.1 | |||
NM_001256674.1 | 2826 | UTR 3 | NP_001243603.1 | |||
NM_001256675.1 | 2826 | UTR 3 | NP_001243604.1 | |||
NM_001256676.1 | 2826 | UTR 3 | NP_001243605.1 | |||
NM_001256677.1 | 2826 | UTR 3 | NP_001243606.1 | |||
NM_001324226.1 | 2826 | UTR 3 | NP_001311155.1 | |||
NM_001324227.1 | 2826 | UTR 3 | NP_001311156.1 | |||
NM_001324228.1 | 2826 | UTR 3 | NP_001311157.1 | |||
NM_001324229.1 | 2826 | UTR 3 | NP_001311158.1 | |||
NM_001324230.1 | 2826 | UTR 3 | NP_001311159.1 | |||
NM_004809.4 | 2826 | UTR 3 | NP_004800.2 | |||
XM_005254792.4 | 2826 | UTR 3 | XP_005254849.1 | |||
XM_006720773.3 | 2826 | UTR 3 | XP_006720836.1 |
Set Membership: |
HapMap Validated |