Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605851 MIM: 616923 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ICMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ICMT - isoprenylcysteine carboxyl methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012405.3 | 4101 | UTR 3 | NP_036537.1 | |||
XM_011541140.2 | 4101 | UTR 3 | XP_011539442.1 |
RNF207 - ring finger protein 207 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207396.2 | 4101 | Intron | NP_997279.2 | |||
XM_011541439.2 | 4101 | Intron | XP_011539741.1 | |||
XM_017001259.1 | 4101 | Intron | XP_016856748.1 |
Set Membership: |
HapMap JSNP Validated |