Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605536 MIM: 615572 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB11FIP5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RAB11FIP5 - RAB11 family interacting protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015470.2 | 6145 | UTR 3 | NP_056285.1 | |||
XM_005264251.3 | 6145 | UTR 3 | XP_005264308.1 | |||
XM_005264252.3 | 6145 | UTR 3 | XP_005264309.1 | |||
XM_005264253.4 | 6145 | Intron | XP_005264310.1 | |||
XM_006711985.3 | 6145 | UTR 3 | XP_006712048.1 | |||
XM_011532753.2 | 6145 | UTR 3 | XP_011531055.1 | |||
XM_011532754.2 | 6145 | Intron | XP_011531056.1 | |||
XM_011532755.2 | 6145 | Intron | XP_011531057.1 | |||
XM_011532756.2 | 6145 | Intron | XP_011531058.1 | |||
XM_017003788.1 | 6145 | UTR 3 | XP_016859277.1 |
SFXN5 - sideroflexin 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |