Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603518 | ||||||||||||||||||||
Literature Links: |
TIA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TIA1 - TIA1 cytotoxic granule-associated RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022037.2 | 1900 | UTR 3 | NP_071320.2 | |||
NM_022173.2 | 1900 | UTR 3 | NP_071505.2 | |||
XM_005264527.1 | 1900 | UTR 3 | XP_005264584.1 | |||
XM_005264528.4 | 1900 | UTR 3 | XP_005264585.1 | |||
XM_005264531.1 | 1900 | UTR 3 | XP_005264588.1 | |||
XM_011533077.1 | 1900 | UTR 3 | XP_011531379.1 | |||
XM_011533078.1 | 1900 | UTR 3 | XP_011531380.1 | |||
XM_011533081.2 | 1900 | UTR 3 | XP_011531383.1 | |||
XM_011533082.2 | 1900 | UTR 3 | XP_011531384.1 | |||
XM_017004789.1 | 1900 | UTR 3 | XP_016860278.1 | |||
XM_017004790.1 | 1900 | UTR 3 | XP_016860279.1 | |||
XM_017004791.1 | 1900 | UTR 3 | XP_016860280.1 | |||
XM_017004792.1 | 1900 | UTR 3 | XP_016860281.1 | |||
XM_017004793.1 | 1900 | UTR 3 | XP_016860282.1 | |||
XM_017004794.1 | 1900 | UTR 3 | XP_016860283.1 | |||
XM_017004795.1 | 1900 | UTR 3 | XP_016860284.1 | |||
XM_017004796.1 | 1900 | UTR 3 | XP_016860285.1 | |||
XM_017004797.1 | 1900 | UTR 3 | XP_016860286.1 | |||
XM_017004798.1 | 1900 | UTR 3 | XP_016860287.1 | |||
XM_017004799.1 | 1900 | UTR 3 | XP_016860288.1 | |||
XM_017004800.1 | 1900 | UTR 3 | XP_016860289.1 |