Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600694 | ||||||||||||||||||||
Literature Links: |
IL6ST PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL6ST - interleukin 6 signal transducer | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190981.1 | 5951 | UTR 3 | NP_001177910.1 | |||
NM_002184.3 | 5951 | UTR 3 | NP_002175.2 | |||
NM_175767.2 | 5951 | UTR 3 | NP_786943.1 | |||
XM_017009441.1 | 5951 | UTR 3 | XP_016864930.1 | |||
XM_017009442.1 | 5951 | Intron | XP_016864931.1 |